missing translation for 'onlineSavingsMsg'
Learn More

Tetra-n-butylammonium acetate, 1.0M aq. soln.

Catalog No. AAB2477314
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2477314 25 g
AAB2477322 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2477314 Supplier Thermo Scientific Chemicals Supplier No. B2477314
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 10534-59-5 | C18H39NO2 | 301.52 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 10534-59-5
Molecular Formula C18H39NO2
Molecular Weight (g/mol) 301.52
MDL Number MFCD00043208
InChI Key MCZDHTKJGDCTAE-UHFFFAOYSA-M
Synonym tetrabutylammonium acetate, 1-butanaminium, n,n,n-tributyl-, acetate, tetra-n-butylammonium acetate, tetrabutylazanium acetate, tetrabutyl ammonium acetate, 1-butanaminium, n,n,n-tributyl-, acetate 1:1, tetrabutylammonium ion acetate, 2-phosphonooxy acrylic acid cyclohexylamine salt 1:1, n,n,n-tributylbutan-1-aminiumacetate, tetrabutylazanium ethanoate
PubChem CID 82707
IUPAC Name tetrabutylazanium;acetate
SMILES CC([O-])=O.CCCC[N+](CCCC)(CCCC)CCCC

Specifications

Density 0.99
Boiling Point 100°C
Refractive Index 1.389
Linear Formula [CH3(CH2)3]4N(O2CCH3)
Quantity 25 g
Beilstein 3599376
Formula Weight 301.51
Concentration or Composition (by Analyte or Components) 1.0M aq. soln.
Chemical Name or Material Tetra-n-butylammonium acetate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 234-101-8
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only