missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sigma Aldrich Tetra(ethylene glycol) diacrylate
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
$330.73 - $330.73
Specifications
| CAS | 17831-71-9 |
|---|---|
| Density | 1.11 g/mL (at 25°C) |
| Molecular Formula | C14H22O7 |
| Linear Formula | (H2C=CHCO2CH2CH2OCH2CH2)2 O |
| Refractive Index | n20/D 1.465 (literature) |
Description
Tetra(ethylene glycol) diacrylateSpecifications
| 17831-71-9 | |
| C14H22O7 | |
| n20/D 1.465 (literature) | |
| TTEGDA |
| 1.11 g/mL (at 25°C) | |
| (H2C=CHCO2CH2CH2OCH2CH2)2 O | |
| MFCD00008630 | |
| 302.32 |