missing translation for 'onlineSavingsMsg'
Learn More

Sigma Aldrich Tetra(ethylene glycol) diacrylate

Catalog No. 3988021L
Encompass
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
3988021L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 3988021L Supplier Sigma Aldrich Supplier No. 3988021L
Only null left

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Tetra(ethylene glycol) diacrylate

Specifications

CAS 17831-71-9
Density 1.11 g/mL (at 25°C)
Molecular Formula C14H22O7
Refractive Index n20/D 1.465 (literature)
Linear Formula (H2C=CHCO2CH2CH2OCH2CH2)2 O
Quantity 1 L
MDL Number MFCD00008630
Synonym TTEGDA
Molecular Weight (g/mol) 302.32