missing translation for 'onlineSavingsMsg'
Learn More

Velp Scientifica™ Test Tubes for Digestion Units

Catalog No. 11392032
Change view
Click to view available options
Diameter (Metric) Outer:
26 mm
42 mm
48 mm
80 mm
For Use With (Equipment):
Digesters DK 20/26, DKL 42/26 and UDK Distillation Units
DK6/48 and UDK series
UDK Series distillation units
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Diameter (Metric) Outer For Use With (Equipment)
11392032 42 mm -
11392034 26 mm Digesters DK 20/26, DKL 42/26 and UDK Distillation Units
11392170 48 mm DK6/48 and UDK series
11392169 80 mm UDK Series distillation units
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 11392032 Supplier Velp Scientifica™ Supplier No. A00000144

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Facilitates the nitrogen analysis and protein determination process. Velp Scientifica™ Test Tubes for Digestion Units are for use with Velp Scientifica™ Kjeldahl Digestion Units.

  • For use in Total Kjeldahl Nitrogen Determination, ammoniacal nitrogen, nitric nitrogen (Devarda), TVBN, sulphites, phenols, volatile acids and cyanides

Specifications

Product Type Test Tubes
Capacity (Metric) 250 mL
Diameter (Metric) Outer 42 mm
Length (Metric) 300 mm
Quantity 3 Tubes