missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Test Tube Racks for MaxQ™ Shaker

Catalog No. 14512118
Change view
Click to view available options
Color:
Blue
Green
Orange
Red
White
Holds:
Tubes (1.5 mL)
Tubes (10 to 13 mm)
Tubes (14 to 16 mm), Centrifuge Tubes (15 mL)
Tubes (17 to 20 mm)
Tubes (21 to 25 mm)
Tubes (26 to 30 mm)
Tubes (26 to 30 mm), Centrifuge Tubes (50 mL)
Array:
3 x 3
3 x 8 array
4 x 10
4 x 4
4 x 5
4 x 6
6 x 12
6 x 6
8 x 12
12 product options available for selection
Product selection table with 12 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Holds Array
14512118 Blue Tubes (1.5 mL) 4 x 6
14512117 Blue Tubes (1.5 mL) 8 x 12
14512108 Red Tubes (10 to 13 mm) 6 x 6
14512107 Red Tubes (10 to 13 mm) 6 x 12
14512110 Orange Tubes (14 to 16 mm), Centrifuge Tubes (15 mL) 6 x 6
14512109 Orange Tubes (14 to 16 mm), Centrifuge Tubes (15 mL) 6 x 12
14512112 White Tubes (17 to 20 mm) 4 x 5
14512111 White Tubes (17 to 20 mm) 4 x 10
14512114 Blue Tubes (21 to 25 mm) 4 x 4
14512113 Blue Tubes (21 to 25 mm) 4 x 10
14512115 Green Tubes (26 to 30 mm) 3 x 8 array
14512116 Green Tubes (26 to 30 mm), Centrifuge Tubes (50 mL) 3 x 3
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 12 options available.
12 options
Catalog No. 14512118 Supplier Thermo Scientific™ Supplier No. 30191

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Mix and match these shaker test tube racks, available in a variety of sizes and arrays, to suit individual needs in the laboratory.

Mix and match Thermo Scientific™ MaxQ™ Shaker Test Tube Racks, available in a variety of sizes and arrays, to suit individual needs in the laboratory. All racks are compatible with MaxQ shaker platforms.

Racks are available in a multitude of sizes, arrays, and colors. Compatible with MaxQ shaker platforms.

Specifications

Description Blue; 1.5 mL; 4 x 6 array
Color Blue
Holds Tubes (1.5 mL)
Positions 24
For Use With (Equipment) MaxQ™ Shaker Platform
Product Type Test Tube Rack
Array 4 x 6