Learn More
Thermo Scientific™ Test Tube Racks for MaxQ™ Shaker
Mix and match these shaker test tube racks, available in a variety of sizes and arrays, to suit individual needs in the laboratory.
Supplier:  Thermo Scientific™ 30181
 View more versions of this product
Catalog No. 14512108
Only null left
Description
Description
Mix and match Thermo Scientific™ MaxQ™ Shaker Test Tube Racks, available in a variety of sizes and arrays, to suit individual needs in the laboratory. All racks are compatible with MaxQ shaker platforms.
Racks are available in a multitude of sizes, arrays, and colors. Compatible with MaxQ shaker platforms.
Specifications
Chemical Identifiers
MaxQ™ Shaker Platform
Test Tube Rack
Specifications
Red; 10 to 13 mm; 6 x 6 array
Tubes (10 to 13 mm)
MaxQ™ Shaker Platform
6 x 6
Red
36
Test Tube Rack
Product Suggestions
Product Suggestions