missing translation for 'onlineSavingsMsg'
Learn More

United Scientific™ Test Tube Drying Rack

Catalog No. S15983
Change view
Click to view available options
Array:
5 x 10
8 x 12
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Array
S15983 5 x 10
S15723 8 x 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S15983 Supplier United Scientific™ Supplier No. 77852

Dry your test tubes without taking up a lot of bench space.

  • These polypropylene racks can conveniently hold inverted tubes for drying and minimize collection or airborne contaminants.
  • Racks can also be used to hold and dry electrophoresis and chromatography plates.
  • Autoclavable

Specifications

Autoclavable Autoclavable
Color Red
Holds Test Tubes
Material Polypropylene
Positions 50
Shape Rectangular
Stackable No
Sterility Non-sterile
For Use With (Equipment) 16 mm Tubes
Includes 6 Racks
Product Type Tube Rack with Drying Pins
Shelves None
Array 5 x 10
Show More Show Less

Warning: Cancer and reproductive harm