Learn More
Eisco™ Test Strips pH 0 to 1.5 Range - High Precision Acid Test Kit, Measures in Increments of 0.3 to 1.5 ppm, 50 Strips
Description
- Precise acid testing: Measures strong acidic solutions from 0 to 1.5 pH in specific increments (0.3, 0.5, 0.8, 1.0, 1.5 ppm)
- Multiple applications: Suitable for laboratory use, industrial testing, and any setting requiring precise acid level monitoring
- Easy to use: Quick dip-and-read test strips with clear instructions to avoid contamination; ideal for accurate readings
- Immediate comparison: Compare the test strip to the color chart immediately after dipping to ensure accuracy
- Strips include a safe handling zone to prevent contact with the sensitive test pad
- Our pH Test Strips provide a reliable method to measure pH levels of strong acidic solutions accurately
- This product caters to environments where precise acidity measurements are crucial, such as chemical processing and scientific research.
- The package includes 100 easy-to-use strips that allow users to quickly determine the acidity of solutions.
- To use, simply dip the strip in the solution, shake off any excess liquid, and compare it to the color chart for results. This product helps streamline the monitoring of pH levels in various settings.
- Please note, this product is intended for testing and monitoring purposes only and is not designed for medical use.
Specifications
Specifications
| pH Range | 0 to 1.5 |
| Type | pH 0-1.5 Test Strips |