Learn More
Stemcell Technologies Inc TeSR™-E5, Size: 1 Kit
Supplier:  Stemcell Technologies Inc STM05916
Encompass
TeSR™-E5 is a defined, serum- and xeno-free medium that is based on the formulation of TeSR™-E8™, but does not contain transforming growth factor (TGF-), basic fibroblast growth factor (bFGF) or insulin. It may be used as a basal medium for differentiation of human embryonic stem (ES) cells and induced pluripotent stem (iPS) cells, or other applications where removal of the above cytokines and insulin is desirable.
Catalog No. NC1520220
Only null left