Learn More
Terumo™ VENOJECT™ II Evacuated Tube Holder Luer Adapter
Catalog No. 22290017
Change view
Click to view available options
Sterility:
Sterile
Catalog No. Sterility
22290017 Sterile
1 options
Catalog No. 22290017 Supplier Terumo™ Supplier No. MN2000T
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Allows multiple draws into evacuated tubes
Specifications
Disposable Single-use
For Use With (Equipment) Standard Tube Holder and SampLok™ Tube Holder
Sterility Sterile
Product Type Luer Adapter
Connectors Male Luer