Learn More
Terumo™ SurGuard™3 Safety Hypodermic Needles, 30G
Catalog No. 22272115
Change view
Click to view available options
Needle Gauge:
30 G
Catalog No. Needle Gauge
22272115 30 G
1 options
Catalog No. 22272115 Supplier Terumo™ Supplier No. SG3 3013
Only null left
With three ways to activate (finger, thumb and hard surface) and a broad range of sizes, SurGuard™3 is the industry's most adaptable safety needle.
  • 3 modes of activation-thumb, finger, or hard surface
  • Separate finger/thumb grips with ridges and nonslip surfaces
  • Hub and needle locks for security post activation
  • Broad range of sizes
  • Firm injection feel
  • Sharpest needles among major brands tested; 10% sharper than similar needles from Becton, Dickinson and Company
  • More comfortable injections for patients
Specifications
Needle Gauge 30 G
Latex Free Yes
Length (English) Needle 0.5 in.
Sterility Sterile