Learn More
Terumo™ Surflo™ ETFE I.V. Catheters
With an ultra-sharp double-bevel introducer needle and medical-grade lubricant, the SURFLO ETFE I.V. catheter allows easier penetration and smoother travel through tissue.
Supplier:  Terumo™ SROX2419CA
Catalog No. 22251491
Only null left
Explore available promotions
Description
Description
  • Ultra-sharp double-bevel introducer needle promotes easier penetration
  • Medical-grade lubricant allows for smoother travel through tissue
  • Preview chamber in clear sure-grip hub gives immediate indication of proper placement
  • Flexible catheter with a large inner diameter assures good blood flow
Specifications
Specifications
ETFE
24 G
27 G ultra-sharp double-bevel needle, medical-grade lubricant and preview chamber in clear sure-grip™ hμb
0.75 in.
50 Box
Sterile
Yellow
Easier Penetration and Smoother Travel Through Tissue
Yes
IV Catheter
Bevel
Product Suggestions
Product Suggestions