Learn More
Terumo™ Hypodermic Syringe with 25 G Needle
Catalog No. 361030871
Change view
Click to view available options
Needle Gauge:
25 G
Catalog No. Needle Gauge
361030871 25 G
1 options
Catalog No. 361030871 Supplier Terumo™ Supplier No. SS03L2516
Only null left
Latex-free, clear barrels with bold scale markings
  • Clear barrels permit quick visual inspection for precise dosage
  • Integrated, smooth plunger stop helps prevent the plunger from being accidentally pulled out
  • Tamper evident blister packs
Specifications
Needle Gauge 25 G
Volume (Metric) 3 mL
Connection Type Luer Lock