Learn More
Terumo™ Hypodermic Syringes
Description
- Clear barrels permit quick visual inspection for precise dosage
- Integrated, smooth plunger stop helps prevent the plunger from being accidentally pulled out
- Tamper evident blister packs
Specifications
Specifications
| Needle Point Number | No Needle Included |
| Volume (Metric) | 10 mL |
| Connection Type | Leur Slip |