Learn More
Terumo™ Hypodermic Syringes
Latex-free, clear barrels with bold scale markings
Supplier: Terumo™ SS10L
Description
- Clear barrels permit quick visual inspection for precise dosage
- Integrated, smooth plunger stop helps prevent the plunger from being accidentally pulled out
- Tamper evident blister packs
Specifications
| No Needle Included | |
| 10 mL | |
| Luer Lock |