Learn More
Terumo™ Hypodermic Syringes
Catalog No. 22272297
Change view
Click to view available options
Volume (Metric):
3 mL
5 mL
10 mL
60 mL
Connection Type:
Leur Slip
Luer Lock
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) Connection Type
22272297 3 mL Leur Slip
22272307 5 mL Luer Lock
22272316 10 mL Leur Slip
22272315 10 mL Luer Lock
22272296 3 mL Luer Lock
22272325 60 mL Luer Lock
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 22272297 Supplier Terumo™ Supplier No. SS03S
Latex-free, clear barrels with bold scale markings
  • Clear barrels permit quick visual inspection for precise dosage
  • Integrated, smooth plunger stop helps prevent the plunger from being accidentally pulled out
  • Tamper evident blister packs
Specifications
Needle Point Number No Needle Included
Volume (Metric) 3 mL
Connection Type Leur Slip