Learn More
Terumo™ Hypodermic Needles
Catalog No. p-184970
Change view
Click to view available options
Needle Gauge:
20 G
21 G
22 G
23 G
25 G
Length (English) Needle:
1 in.
Needle Point Style:
Thin Wall
Ultra-thin Wall
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Needle Gauge Length (English) Needle Needle Point Style
22272284 22 G 1 in. Ultra-thin Wall
22272286 23 G 1 in. Ultra-thin Wall
22272282 21 G 1 in. Ultra-thin Wall
22272280 20 G 1 in. Ultra-thin Wall
22272288 25 G 1 in. Thin Wall
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 22272284 Supplier Terumo™ Supplier No. TCNN2225R
Only null left
Provides ultra sharp needles, which translate into greater comfort for the patient.
  • Double bevel design
  • Precision manufactured ultrathin wall
Specifications
Needle Gauge 22 G
Latex Free Yes
Length (English) Needle 1 in.
Product Type Double Bevel Design
Needle Point Style Ultra-thin Wall