missing translation for 'onlineSavingsMsg'
Learn More

tert-Pentylamine, 98%

Catalog No. p-7027687
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2463914 25 g
AAB2463922 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2463914 Supplier Thermo Scientific Chemicals Supplier No. B2463914
Only null left

CAS: 594-39-8 | C5H13N | 87.166 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 594-39-8
Molecular Formula C5H13N
Molecular Weight (g/mol) 87.166
MDL Number MFCD00008056
InChI Key GELMWIVBBPAMIO-UHFFFAOYSA-N
Synonym tert-amylamine, tert-pentylamine, 2-butanamine, 2-methyl, t-amylamine, 1,1-dimethylpropylamine, 2-methyl-2-butylamine, 2-amino-2-methylbutane, 2-methyl-butan-2-amine, 1,1-dimethyl-1-propylamine, t-pentylamine
PubChem CID 68986
ChEBI CHEBI:84236
IUPAC Name 2-methylbutan-2-amine
SMILES CCC(C)(C)N

Specifications

Density 0.746
Boiling Point 76°C to 78°C
Flash Point −1°C (30°F)
Odor Amine-like
Refractive Index 1.3995
Quantity 25 g
UN Number UN1106
Sensitivity Air Sensitive
Formula Weight 87.17
Percent Purity 98%
Chemical Name or Material tert-Pentylamine
Show More Show Less
Hazard Category H225-H302+H312+H332-H314
Hazard Statement GHS H Statement
H225-H314-H318-H302-H312-H332
Highly flammable liquid and vapour.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Precautionary Statement P210-P233-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: AMYLAMINES
EINECSNumber 209-839-9
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only