missing translation for 'onlineSavingsMsg'
Learn More
Learn More
tert-Nonyl Mercaptan (mixture) 97.0+%, TCI America™
$46.60 - $210.63
Chemical Identifiers
| CAS | 25360-10-5 |
|---|---|
| Molecular Formula | C9H20S |
| Molecular Weight (g/mol) | 160.32 |
| MDL Number | MFCD00059963 |
| InChI Key | MPBLPZLNKKGCGP-UHFFFAOYSA-N |
| PubChem CID | 520196 |
| IUPAC Name | 2-methyloctane-2-thiol |
| SMILES | CCCCCCC(C)(C)S |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
N029925ML
|
TCI America
N029925ML |
25 mL |
Each for $46.60
|
|
|||||
|
N0299500ML
|
TCI America
N0299500ML |
500 mL |
Each for $210.63
|
|
|||||
Chemical Identifiers
| 25360-10-5 | |
| 160.32 | |
| MPBLPZLNKKGCGP-UHFFFAOYSA-N | |
| 2-methyloctane-2-thiol |
| C9H20S | |
| MFCD00059963 | |
| 520196 | |
| CCCCCCC(C)(C)S |
Specifications
| 25360-10-5 | |
| C9H20S | |
| 25 mL | |
| CCCCCCC(C)(C)S | |
| 160.32 | |
| 160.32 | |
| Liquid |
| Yellow | |
| MFCD00059963 | |
| MPBLPZLNKKGCGP-UHFFFAOYSA-N | |
| 2-methyloctane-2-thiol | |
| 520196 | |
| ≥97.0% (T) | |
| tert-Nonyl Mercaptan (mixture) |
Safety and Handling
EINECSNumber : (2)-0464
TSCA : Yes