missing translation for 'onlineSavingsMsg'
Learn More

tert-Butylsulfinamide 97.0+%, TCI America™

Catalog No. p-7119845
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B29261G 1 g
B29265G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B29261G Supplier TCI America Supplier No. B29261G
Only null left

Chemical Identifiers

CAS 146374-27-8
Molecular Formula C4H11NOS
Molecular Weight (g/mol) 121.20
MDL Number MFCD01863616,MFCD05861480
InChI Key CESUXLKAADQNTB-UHFFFAOYNA-N
Synonym 2-methyl-2-propanesulfinamide, tert-butanesulfinamide, t-butylsulfinamide, tert-butylsulfinamide, 1,1-dimethylethylsulfinamide, 2-propanesulfinamide, 2-methyl, 2-methyl-propane-2-sulfinic acid amide, tert-butylsulphinamide, unii-i85yc2za8o, i85yc2za8o
PubChem CID 3382465
IUPAC Name 2-methylpropane-2-sulfinamide
SMILES CC(C)(C)S(N)=O

Specifications

Melting Point 107°C
Color White
Quantity 1 g
Formula Weight 121.20
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material tert-Butylsulfinamide
TSCA No
Recommended Storage Refrigerator