missing translation for 'onlineSavingsMsg'
Learn More

tert-Butylsulfinamide, 98%

Catalog No. AC426830050
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC426830050 5 g Glass bottle
AC426830250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC426830050 Supplier Thermo Scientific Chemicals Supplier No. 426830050
Only null left

CAS: 146374-27-8 | C4H11NOS | 121.20 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 146374-27-8
Molecular Formula C4H11NOS
Molecular Weight (g/mol) 121.20
MDL Number MFCD01863616,MFCD05861480
InChI Key CESUXLKAADQNTB-UHFFFAOYNA-N
Synonym 2-methyl-2-propanesulfinamide, tert-butanesulfinamide, t-butylsulfinamide, tert-butylsulfinamide, 1,1-dimethylethylsulfinamide, 2-propanesulfinamide, 2-methyl, 2-methyl-propane-2-sulfinic acid amide, tert-butylsulphinamide, unii-i85yc2za8o, i85yc2za8o
PubChem CID 3382465
IUPAC Name 2-methylpropane-2-sulfinamide
SMILES CC(C)(C)S(N)=O

Specifications

Melting Point 97.0°C to 101.0°C
Color Tan to White
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula (CH3)3CS(O)NH2
Quantity 5 g
Merck Index 15,3607
Formula Weight 121.2
Percent Purity 98%
Physical Form Crystals or Powder
Chemical Name or Material tert-Butylsulfinamide
Show More Show Less

RUO – Research Use Only