missing translation for 'onlineSavingsMsg'
Learn More

tert-Butylmalononitrile 95.0+%, TCI America™

Catalog No. B15845G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B15845G 5 g
B158425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B15845G Supplier TCI America Supplier No. B15845G
Only null left

Chemical Identifiers

CAS 4210-60-0
Molecular Formula C7H10N2
Molecular Weight (g/mol) 122.17
MDL Number MFCD00191326
InChI Key TUJPHMQZSDKRSL-UHFFFAOYSA-N
PubChem CID 138154
IUPAC Name 2-tert-butylpropanedinitrile
SMILES CC(C)(C)C(C#N)C#N

Specifications

Color White-Yellow
Quantity 5 g
UN Number 3439
Formula Weight 122.17
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material tert-Butylmalononitrile