missing translation for 'onlineSavingsMsg'
Learn More

tert-Butylhydrazine Hydrochloride 98.0+%, TCI America™

Catalog No. p-7118911
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B119525G 25 g
B1195100G 100 g
B1195500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. B119525G Supplier TCI America Supplier No. B119525G
Only null left

Chemical Identifiers

CAS 7400-27-3
Molecular Formula C4H13ClN2
Molecular Weight (g/mol) 124.61
MDL Number MFCD00012947
InChI Key DDPWVABNMBRBFI-UHFFFAOYSA-N
Synonym tert-butylhydrazine hydrochloride, hydrazine, 1,1-dimethylethyl-, monohydrochloride, tert-butylhydrazinium chloride, tert-butylhydrazine monohydrochloride, t-butylhydrazine hydrochloride, hydrazine, 1,1-dimethylethyl-, hydrochloride 1:1, 1,1-dimethylethyl hydrazine monohydrochloride, 2-tert-butylhydrazinium chloride, t-butylhydrazine.hcl, t-bunhnh2.hcl
PubChem CID 81889
IUPAC Name hydrogen tert-butylhydrazine chloride
SMILES [H+].[Cl-].CC(C)(C)NN

Specifications

Color White-Yellow
Quantity 25 g
Formula Weight 124.61
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material tert-Butylhydrazine Hydrochloride
RTECSNumber MV2841000
TSCA Yes