missing translation for 'onlineSavingsMsg'
Learn More

tert-Butylhydrazine hydrochloride, 98%

Catalog No. p-7027258
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2387614 25 g
AAB2387622 100 g
AAB2387636 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2387614 Supplier Thermo Scientific Chemicals Supplier No. B2387614

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 7400-27-3 | C4H13ClN2 | 124.61 g/mol

It is used in organic synthesis for aromatic nucleophilic N-N exchange reaction. tert-Butylhydrazine Hydrochloride is also used in the preparation of insect growth regulator. tert-Butylhydrazine hydrochloride was used in the synthesis of diphosphinohydrazines, 5-amino-1-tert-butyl-3-(p-chlorophenyl)-4-cyanopyrazole and 1,3,4,5-tetrasubstituted pyrazole derivatives.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used in organic synthesis for aromatic nucleophilic N-N exchange reaction. tert-Butylhydrazine Hydrochloride is also used in the preparation of insect growth regulator. tert-Butylhydrazine hydrochloride was used in the synthesis of diphosphinohydrazines, 5-amino-1-tert-butyl-3-(p-chlorophenyl)-4-cyanopyrazole and 1,3,4,5-tetrasubstituted pyrazole derivatives.

Solubility
Solubility in water (almost transparency). Soluble in DMSO and methanol.

Notes
Hygroscopic. Store away from water/moisture and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas. Protect from humidity and water.

Chemical Identifiers

CAS 7400-27-3
Molecular Formula C4H13ClN2
Molecular Weight (g/mol) 124.61
MDL Number MFCD00012947
InChI Key DDPWVABNMBRBFI-UHFFFAOYSA-N
Synonym tert-butylhydrazine hydrochloride, hydrazine, 1,1-dimethylethyl-, monohydrochloride, tert-butylhydrazinium chloride, tert-butylhydrazine monohydrochloride, t-butylhydrazine hydrochloride, hydrazine, 1,1-dimethylethyl-, hydrochloride 1:1, 1,1-dimethylethyl hydrazine monohydrochloride, 2-tert-butylhydrazinium chloride, t-butylhydrazine.hcl, t-bunhnh2.hcl
PubChem CID 81889
SMILES [H+].[Cl-].CC(C)(C)NN

Specifications

Melting Point ∼192°C (decomposition)
Quantity 25 g
Beilstein 3651224
IUPAC Name tert-butylhydrazine;hydrochloride
Formula Weight 124.61
Percent Purity 98%
Chemical Name or Material tert-Butylhydrazine hydrochloride
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 231-001-6
RTECSNumber MV2841000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only