missing translation for 'onlineSavingsMsg'
Learn More
Learn More
tert-Butylferrocene 97.0+%, TCI America™
$261.82 - $787.39
Chemical Identifiers
| CAS | 1316-98-9 |
|---|---|
| Molecular Formula | C14H18Fe |
| MDL Number | MFCD00058729 |
Chemical Identifiers
| 1316-98-9 | |
| C14H18Fe | |
| MFCD00058729 |
Specifications
| 1316-98-9 | |
| MFCD00058729 | |
| 242.14 | |
| Liquid |
| C14H18Fe | |
| 1 g | |
| ≥97.0% (GC) | |
| tert-Butylferrocene |
Safety and Handling
TSCA : No