missing translation for 'onlineSavingsMsg'
Learn More

tert-Butylferrocene 97.0+%, TCI America™

Catalog No. B20635G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B20635G 5 g
B20631G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B20635G Supplier TCI America Supplier No. B20635G

Chemical Identifiers

CAS 1316-98-9
Molecular Formula C14H18Fe
MDL Number MFCD00058729

Specifications

Quantity 5 g
Formula Weight 242.14
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material tert-Butylferrocene