missing translation for 'onlineSavingsMsg'
Learn More
Learn More
tert-Butylferrocene 97.0+%, TCI America™
Specifications
Specifications
| Quantity | 1 g |
| Formula Weight | 242.14 |
| Percent Purity | ≥97.0% (GC) |
| Physical Form | Liquid |
| Chemical Name or Material | tert-Butylferrocene |
Safety and Handling
| TSCA | No |