missing translation for 'onlineSavingsMsg'
Learn More

tert-Butylferrocene, 98%, Thermo Scientific Chemicals

Catalog No. AA3940506
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3940506 5 g
AA3940503 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA3940506 Supplier Thermo Scientific Chemicals Supplier No. 03940506

It finds its application in microwave-enhanced electrochemical processes in micellar surfactant media.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1316-98-9
MDL Number MFCD00058729

Specifications

Density 1.201 g/mL
Boiling Point 96°C (1 mmHg)
Refractive Index 1.579
Quantity 5 g
Solubility Information Not miscible or difficult to mix in water.
Formula Weight 242.15
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material tert-Butylferrocene
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only