Learn More
tert-Butyldiphenylchlorosilane, 97%
Description
tert-Butyldiphenylchlorosilane acts as a silylating reagent used to protect alcohols and in the preparation of silyl ethers. It plays a major role as a raw material and a precursor in organic synthesis and pharmaceuticals. It is also used for the synthesis of interphenylene phenyloxazoles which can be used for the treatment of circulatory disorders, angina and stroke.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
tert-Butyldiphenylchlorosilane acts as a silylating reagent used to protect alcohols and in the preparation of silyl ethers. It plays a major role as a raw material and a precursor in organic synthesis and pharmaceuticals. It is also used for the synthesis of interphenylene phenyloxazoles which can be used for the treatment of circulatory disorders, angina and stroke.
Solubility
Miscible with chloroform and ethyl acetate. Immiscible with water.
Notes
Moisture sensitive. Incompatible with strong oxidizing agents, water and bases.
Specifications
Specifications
| Boiling Point | 90°C to 92°C (0.01mmHg) |
| Quantity | 250 g |
| Flash Point | 112°C (233°F) |
| Refractive Index | 1.568 |
| Sensitivity | Moisture sensitive |
| Density | 1.072 |
| UN Number | UN2987 |
| Beilstein | 644023 |
| Solubility Information | Miscible with chloroform and ethyl acetate. Immiscible with water. |
| Formula Weight | 274.87 |
| Show More |
Safety and Handling
| Hazard Category | H314-H335 |
| Hazard Statement | GHS H Statement H314-H318 Causes severe skin burns and eye damage. Causes serious eye damage. |
| Precautionary Statement | P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P312-P363-P501c |
| DOTInformation | Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CHLOROSILANES, CORROSIVE, N.O.S. |
| EINECSNumber | 261-282-0 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures; Store under Nitrogen |
RUO – Research Use Only