missing translation for 'onlineSavingsMsg'
Learn More

(tert-Butyldimethylsilyloxy)malononitrile 93.0+%, TCI America™

Catalog No. B5370200MG
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B5370200MG 200 mg
B53701G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B5370200MG Supplier TCI America Supplier No. B5370200MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 128302-78-3
Molecular Formula C9H16N2OSi
Molecular Weight (g/mol) 196.325
MDL Number MFCD27942977
InChI Key AIQDCGCDLHCZHB-UHFFFAOYSA-N
Synonym (tert-Butyldimethylsilyloxy)propanedinitrile, MAC-TBS Reagent
PubChem CID 10856410
IUPAC Name 2-[tert-butyl(dimethyl)silyl]oxypropanedinitrile
SMILES CC(C)(C)[Si](C)(C)OC(C#N)C#N

Specifications

Color Yellow
Quantity 200 mg
UN Number 3276
Formula Weight 196.33
Percent Purity ≥93.0% (GC)
Physical Form Liquid
Chemical Name or Material (tert-Butyldimethylsilyloxy)malononitrile
TSCA No
Recommended Storage Refrigerator