missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyldimethylsilyl Trifluoromethanesulfonate 98.0+%, TCI America™

Catalog No. p-7136819
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T15255G 5 g
T152525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T15255G Supplier TCI America Supplier No. T15255G
Only null left

Chemical Identifiers

CAS 69739-34-0
Molecular Formula C7H15F3O3SSi
Molecular Weight (g/mol) 264.33
MDL Number MFCD00000405
InChI Key WLLIXJBWWFGEHT-UHFFFAOYSA-N
Synonym tert-butyldimethylsilyl trifluoromethanesulfonate, trifluoromethanesulfonic acid tert-butyldimethylsilyl ester, tbdms triflate, t-butyldimethylsilyl triflate, tert-butyldimethylsilyl trifluoromethanesulphonate, tbdms-otf, tert-butyldimethylsilyl triflate, silane tbm2 triflate, t-butyldimethylsilyltrifluoromethanesulfonate, t-butylmethylsilyl triflate
PubChem CID 123564
IUPAC Name tert-butyldimethylsilyl trifluoromethanesulfonate
SMILES CC(C)(C)[Si](C)(C)OS(=O)(=O)C(F)(F)F

Specifications

Color Yellow
Quantity 5 g
UN Number 2924
Formula Weight 264.33
Percent Purity ≥98.0% (T)
Physical Form Liquid
Chemical Name or Material tert-Butyldimethylsilyl Trifluoromethanesulfonate