missing translation for 'onlineSavingsMsg'
Learn More
Learn More
tert-Butyldimethylsilyl Trifluoromethanesulfonate 98.0+%, TCI America™
Specifications
Specifications
| Color | Yellow |
| Quantity | 5 g |
| UN Number | 2924 |
| Formula Weight | 264.33 |
| Percent Purity | ≥98.0% (T) |
| Grade | Not Available |
| Physical Form | Liquid |
| Chemical Name or Material | tert-Butyldimethylsilyl Trifluoromethanesulfonate |
Safety and Handling
| TSCA | No |