missing translation for 'onlineSavingsMsg'
Learn More

(tert-Butyldimethylsiloxy)acetaldehyde, 90%

Catalog No. AC397510010
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC397510010 1 g Glass bottle
AC397510050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC397510010 Supplier Thermo Scientific Chemicals Supplier No. 397510010

CAS: 102191-92-4 | C8H18O2Si | 174.32 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 102191-92-4
Molecular Formula C8H18O2Si
Molecular Weight (g/mol) 174.32
MDL Number MFCD01321229
InChI Key MEBFFOKESLAUSJ-UHFFFAOYSA-N
Synonym tert-butyldimethylsilyloxy acetaldehyde, 2-tert-butyldimethylsilyl oxy acetaldehyde, 2-tert-butyldimethylsilyloxy acetaldehyde, tert-butyldimethylsiloxy acetaldehyde, t-butyldimethylsilyloxyacetaldehyde, 2-tert-butyl dimethyl silyl oxyacetaldehyde, acetaldehyde, 1,1-dimethylethyl dimethylsilyl oxy, pubchem20197
PubChem CID 4187788
SMILES CC(C)(C)[Si](C)(C)OCC=O

Specifications

CAS Min % 88.0
CAS Max % 100.0
Color Gray to White
Density 0.9150g/mL
Boiling Point 156°C to 157°C
Flash Point 60°C
Infrared Spectrum Authentic
Assay Percent Range 88% min. (GC)
Packaging Glass bottle
Refractive Index 1.4310 to 1.4330 (20°C, 589nm)
Linear Formula (CH3)3CSi(CH3)2OCH2C(O)H
Quantity 1 g
Specific Gravity 0.915
IUPAC Name 2-[tert-butyl(dimethyl)silyl]oxyacetaldehyde
Formula Weight 174.31
Percent Purity 90%
Physical Form Powder
Chemical Name or Material (tert-Butyldimethylsiloxy)acetaldehyde
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Flammable liquid and vapor.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only