missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyldimethylsilanol 95.0+%, TCI America™

Catalog No. p-7124099
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B54995ML 5 mL
B549925ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B54995ML Supplier TCI America Supplier No. B54995ML
Only null left

Chemical Identifiers

CAS 18173-64-3
Molecular Formula C6H16OSi
Molecular Weight (g/mol) 132.278
MDL Number MFCD00010673
InChI Key FGWRMMTYIZKYMA-UHFFFAOYSA-N
PubChem CID 554527
IUPAC Name tert-butyl-hydroxy-dimethylsilane
SMILES CC(C)(C)[Si](C)(C)O

Specifications

Color Yellow
Boiling Point 139°C
Quantity 5 mL
UN Number 1987
Formula Weight 132.28
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material tert-Butyldimethylsilanol