missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyldimethylchlorosilane 98.0+%, TCI America™

Supplier:  TCI America B099525G

 View more versions of this product

Catalog No. B099525G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

tert-Butyldimethylchlorosilane [tert-Butyldimethylsilylating Agent]
88°C
125°C
MFCD00000501
2925
BCNZYOJHNLTNEZ-UHFFFAOYSA-N
tert-butyl-chloro-dimethylsilane
28928
150.72
Crystalline Powder
18162-48-6
White
C6H15ClSi
25 g
tert-butyldimethylsilyl chloride, tert-butyldimethylchlorosilane, t-butyldimethylchlorosilane, tert-butylchlorodimethylsilane, tbdms chloride, tert-butyl chloro dimethylsilane, chloro-tert-butyldimethylsilane, silane, chloro 1,1-dimethylethyl dimethyl, tbscl, t-butyldimethylsilyl chloride
CC(C)(C)[Si](C)(C)Cl
150.721
CHEBI:85071
≥98.0% (GC)

Chemical Identifiers

18162-48-6
150.721
BCNZYOJHNLTNEZ-UHFFFAOYSA-N
28928
tert-butyl-chloro-dimethylsilane
C6H15ClSi
MFCD00000501
tert-butyldimethylsilyl chloride, tert-butyldimethylchlorosilane, t-butyldimethylchlorosilane, tert-butylchlorodimethylsilane, tbdms chloride, tert-butyl chloro dimethylsilane, chloro-tert-butyldimethylsilane, silane, chloro 1,1-dimethylethyl dimethyl, tbscl, t-butyldimethylsilyl chloride
CHEBI:85071
CC(C)(C)[Si](C)(C)Cl
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-2041

RTECSNumber : VV2000000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions