missing translation for 'onlineSavingsMsg'
Learn More
Learn More
tert-Butyldimethylchlorosilane 98.0+%, TCI America™
Supplier: TCI America B0995100G
Specifications
| tert-Butyldimethylchlorosilane [tert-Butyldimethylsilylating Agent] | |
| 88°C | |
| 125°C | |
| MFCD00000501 | |
| 2925 | |
| BCNZYOJHNLTNEZ-UHFFFAOYSA-N | |
| tert-butyl-chloro-dimethylsilane | |
| 28928 | |
| 150.72 | |
| Crystalline Powder |
| 18162-48-6 | |
| White | |
| C6H15ClSi | |
| 100 g | |
| tert-butyldimethylsilyl chloride, tert-butyldimethylchlorosilane, t-butyldimethylchlorosilane, tert-butylchlorodimethylsilane, tbdms chloride, tert-butyl chloro dimethylsilane, chloro-tert-butyldimethylsilane, silane, chloro 1,1-dimethylethyl dimethyl, tbscl, t-butyldimethylsilyl chloride | |
| CC(C)(C)[Si](C)(C)Cl | |
| 150.721 | |
| CHEBI:85071 | |
| ≥98.0% (GC) |
Chemical Identifiers
| 18162-48-6 | |
| 150.721 | |
| BCNZYOJHNLTNEZ-UHFFFAOYSA-N | |
| 28928 | |
| tert-butyl-chloro-dimethylsilane |
| C6H15ClSi | |
| MFCD00000501 | |
| tert-butyldimethylsilyl chloride, tert-butyldimethylchlorosilane, t-butyldimethylchlorosilane, tert-butylchlorodimethylsilane, tbdms chloride, tert-butyl chloro dimethylsilane, chloro-tert-butyldimethylsilane, silane, chloro 1,1-dimethylethyl dimethyl, tbscl, t-butyldimethylsilyl chloride | |
| CHEBI:85071 | |
| CC(C)(C)[Si](C)(C)Cl |
Safety and Handling
EINECSNumber : (2)-2041
RTECSNumber : VV2000000
TSCA : Yes