missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyldichlorophosphine, Thermo Scientific Chemicals

$102.97 - $102.97

Chemical Identifiers

CAS 25979-07-1
Molecular Formula C4H9Cl2P
Molecular Weight (g/mol) 158.98
MDL Number MFCD00013615
InChI Key NMJASRUOIRRDSX-UHFFFAOYSA-N
Synonym tert-butyldichlorophosphine, t-butyldichlorophosphine, tert-c4h9pcl2, tert-butyl dichloro phosphane, phosphonous dichloride, 1,1-dimethylethyl, snpladsptpdwuup@, pubchem6471, acmc-1cmec, tert-butyl dichloro phosphine, tert-butylphosphine dichloride
PubChem CID 117690
IUPAC Name tert-butyl(dichloro)phosphane
SMILES CC(C)(C)P(Cl)Cl
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC300530025
SDS View Documents
Thermo Scientific Chemicals
300530025
2.5 g Glass bottle
Each for $102.97
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

25979-07-1
158.98
NMJASRUOIRRDSX-UHFFFAOYSA-N
117690
CC(C)(C)P(Cl)Cl
C4H9Cl2P
MFCD00013615
tert-butyldichlorophosphine, t-butyldichlorophosphine, tert-c4h9pcl2, tert-butyl dichloro phosphane, phosphonous dichloride, 1,1-dimethylethyl, snpladsptpdwuup@, pubchem6471, acmc-1cmec, tert-butyl dichloro phosphine, tert-butylphosphine dichloride
tert-butyl(dichloro)phosphane

Specifications

25979-07-1
White to Yellow
Authentic
C4H9Cl2P
MFCD00013615
tert-butyldichlorophosphine, t-butyldichlorophosphine, tert-c4h9pcl2, tert-butyl dichloro phosphane, phosphonous dichloride, 1,1-dimethylethyl, snpladsptpdwuup@, pubchem6471, acmc-1cmec, tert-butyl dichloro phosphine, tert-butylphosphine dichloride
NMJASRUOIRRDSX-UHFFFAOYSA-N
tert-butyl(dichloro)phosphane
117690
Crystalline Low Melting Solid
49.0°C
142.0°C to 150.0°C
Glass bottle
(CH3)3CPCl2
2.5 g
Solubility in water: reacts. Other solubilities: soluble in n-pentane,n-hexane,ether,toluene,,thf
CC(C)(C)P(Cl)Cl
158.98
158.98
tert-Butyldichlorophosphine
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Contact with water liberates toxic gas.
Reacts violently with water.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 247-386-9

SDS
Documents

Documents

RUO – Research Use Only