missing translation for 'onlineSavingsMsg'
Learn More

tert-Butylcyclohexane 99.0+%, TCI America™

Catalog No. p-7118691
Change view
Click to view available options
Quantity:
100 mL
25 mL
Catalog No. Quantity
B082325ML 25 mL
B0823100ML 100 mL
2 options

Catalog No. B082325ML

Supplier: TCI America B082325ML

Only null left

Chemical Identifiers

CAS 3178-22-1
Molecular Formula C10H20
Molecular Weight (g/mol) 140.27
MDL Number MFCD00001471
InChI Key XTVMZZBLCLWBPM-UHFFFAOYSA-N
Synonym t-butylcyclohexane, cyclohexane, tert-butyl, 1-tert-butylcyclohexane, cyclohexane, 1,1-dimethylethyl, unii-0rgt19ucq6, 1,1-dimethylethyl cyclohexane, 0rgt19ucq6, t-butyl cyclohexane, 4-t-butylcyclohexane, tert-butyl-cyclohexane
PubChem CID 18508
IUPAC Name tert-butylcyclohexane
SMILES CC(C)(C)C1CCCCC1

Specifications

Color Colorless
Boiling Point 171°C
Quantity 25 mL
UN Number 3295
Formula Weight 140.27
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material tert-Butylcyclohexane
RTECSNumber GU9384375
TSCA No