missing translation for 'onlineSavingsMsg'
Learn More

tert-Butylcyclohexane 99.0+%, TCI America™

Catalog No. B082325ML
Change view
Click to view available options
Quantity:
25 mL
100 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B082325ML 25 mL
B0823100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B082325ML Supplier TCI America Supplier No. B082325ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 3178-22-1
Molecular Formula C10H20
Molecular Weight (g/mol) 140.27
MDL Number MFCD00001471
InChI Key XTVMZZBLCLWBPM-UHFFFAOYSA-N
Synonym t-butylcyclohexane, cyclohexane, tert-butyl, 1-tert-butylcyclohexane, cyclohexane, 1,1-dimethylethyl, unii-0rgt19ucq6, 1,1-dimethylethyl cyclohexane, 0rgt19ucq6, t-butyl cyclohexane, 4-t-butylcyclohexane, tert-butyl-cyclohexane
PubChem CID 18508
IUPAC Name tert-butylcyclohexane
SMILES CC(C)(C)C1CCCCC1

Specifications

Color Colorless
Boiling Point 171°C
Quantity 25 mL
UN Number 3295
Formula Weight 140.27
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material tert-Butylcyclohexane
RTECSNumber GU9384375
TSCA No