missing translation for 'onlineSavingsMsg'
Learn More

tert-Butylcyclohexane, 99+%

CAS: 3178-22-1 | C10H20 | 140.27 g/mol

Supplier:  Thermo Scientific Chemicals B2412614

 View more versions of this product

Catalog No. AAB2412614


Only null left
Explore available promotions

Description

Description

Used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used as pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
Specifications

Specifications

tert-Butylcyclohexane
-41°C
171°C
C10H20
MFCD00001471
UN3295
Insoluble in water.
CC(C)(C)C1CCCCC1
140.27
140.27
3178-22-1
0.83
40°C (104°F)
1.447
25 g
t-butylcyclohexane, cyclohexane, tert-butyl, 1-tert-butylcyclohexane, cyclohexane, 1,1-dimethylethyl, unii-0rgt19ucq6, 1,1-dimethylethyl cyclohexane, 0rgt19ucq6, t-butyl cyclohexane, 4-t-butylcyclohexane, tert-butyl-cyclohexane
XTVMZZBLCLWBPM-UHFFFAOYSA-N
tert-butylcyclohexane
18508
≥99%

Chemical Identifiers

3178-22-1
140.27
XTVMZZBLCLWBPM-UHFFFAOYSA-N
18508
CC(C)(C)C1CCCCC1
C10H20
MFCD00001471
t-butylcyclohexane, cyclohexane, tert-butyl, 1-tert-butylcyclohexane, cyclohexane, 1,1-dimethylethyl, unii-0rgt19ucq6, 1,1-dimethylethyl cyclohexane, 0rgt19ucq6, t-butyl cyclohexane, 4-t-butylcyclohexane, tert-butyl-cyclohexane
tert-butylcyclohexane
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H226
Flammable liquid and vapour.

P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c

H226

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: HYDROCARBONS, LIQUID, N.O.S.

EINECSNumber : 221-652-4

RTECSNumber : GU9384375

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only