missing translation for 'onlineSavingsMsg'
Learn More

tert-Butylchlorodiphenylsilane, 98%, AcroSeal™

Catalog No. AC430921000
Change view
Click to view available options
Quantity:
100 mL
Packaging:
AcroSeal™ Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC430921000 100 mL AcroSeal™ Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC430921000 Supplier Thermo Scientific Chemicals Supplier No. 430921000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 58479-61-1 | C16H19ClSi | 274.86 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 58479-61-1
Molecular Formula C16H19ClSi
Molecular Weight (g/mol) 274.86
MDL Number MFCD00000497
InChI Key MHYGQXWCZAYSLJ-UHFFFAOYSA-N
Synonym tert-butylchlorodiphenylsilane, tert-butyldiphenylchlorosilane, t-butyldiphenylchlorosilane, silane, chloro 1,1-dimethylethyl diphenyl, tert-butyl chloro diphenylsilane, t-butylchlorodiphenylsilane, tbdpscl, t-butyldiphenylsilyl chloride, tert-butyldiphenyl chlorosilane, unii-3beu48ui4e
PubChem CID 94078
IUPAC Name tert-butyl-chloro-diphenylsilane
SMILES CC(C)(C)[Si](C1=CC=CC=C1)(C2=CC=CC=C2)Cl

Specifications

Color Brown or Colorless to Yellow
Density 1.0600g/mL
Boiling Point 135.0°C (0.7 mmHg)
Flash Point 149°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging AcroSeal™ Glass Bottle
Refractive Index 1.5665 to 1.5685
Linear Formula (CH3)3CSi(C6H5)2Cl
Quantity 100 mL
Specific Gravity 1.06
Merck Index 15,1568
Solubility Information Solubility in water: practically insoluble. Other solubilities: soluble in non-protic solvents,e.g. aliphatic,or aromatic (chlorinated) hydrocarbons
Viscosity 16 mPa.s (25°C)
Formula Weight 274.86
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material tert-Butylchlorodiphenylsilane
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF INHALED: Remove to fresh air and keep at
EINECSNumber 261-282-
Recommended Storage May darken during storage

RUO – Research Use Only