missing translation for 'onlineSavingsMsg'
Learn More

tert-Butylamine 98.0+%, TCI America™

Catalog No. B070925ML
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B070925ML 25 mL
B0709100ML 100 mL
B0709500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. B070925ML Supplier TCI America Supplier No. B070925ML

Chemical Identifiers

CAS 75-64-9
Molecular Formula C4H11N
Molecular Weight (g/mol) 73.139
MDL Number MFCD00008050
InChI Key YBRBMKDOPFTVDT-UHFFFAOYSA-N
Synonym tert-butylamine, erbumine, 2-amino-2-methylpropane, t-butylamine, 2-propanamine, 2-methyl, trimethylaminomethane, 1,1-dimethylethylamine, 2-aminoisobutane, 1,1-dimethylethanamine, butylamine, tert
PubChem CID 6385
ChEBI CHEBI:44639
IUPAC Name 2-methylpropan-2-amine
SMILES CC(C)(C)N

Specifications

Color Colorless
Boiling Point 46°C
Quantity 25 mL
UN Number 3286
Formula Weight 73.14
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material tert-Butylamine
EINECSNumber (2)-0132
RTECSNumber EO3330000
TSCA Yes