missing translation for 'onlineSavingsMsg'
Learn More

tert-Butylamine Hydroiodide 97.0+%, TCI America™

Catalog No. B44341G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B44341G 1 g
B44345G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B44341G Supplier TCI America Supplier No. B44341G

Chemical Identifiers

CAS 39557-45-4
Molecular Formula C4H12IN
Molecular Weight (g/mol) 201.051
InChI Key NLJDBTZLVTWXRG-UHFFFAOYSA-N
Synonym tert-Butylammonium Iodide
PubChem CID 91972147
IUPAC Name 2-methylpropan-2-amine;hydroiodide
SMILES CC(C)(C)N.I

Specifications

Melting Point 221°C
Color White
Quantity 1 g
Formula Weight 201.05
Percent Purity ≥97.0% (N,T)
Physical Form Crystalline Powder
Chemical Name or Material tert-Butylamine Hydroiodide