missing translation for 'onlineSavingsMsg'
Learn More
Learn More
tert-Butylamine Hydrobromide 98.0+%, TCI America™
$46.20 - $136.69
Chemical Identifiers
| CAS | 60469-70-7 |
|---|---|
| Molecular Formula | C4H12BrN |
| Molecular Weight (g/mol) | 154.05 |
| MDL Number | MFCD00674188 |
| InChI Key | CQKAPARXKPTKBK-UHFFFAOYSA-N |
| Synonym | tert-Butylammonium Bromide |
| PubChem CID | 3085186 |
| IUPAC Name | hydrogen 2-methylpropan-2-amine bromide |
| SMILES | [H+].[Br-].CC(C)(C)N |
Chemical Identifiers
| 60469-70-7 | |
| 154.05 | |
| CQKAPARXKPTKBK-UHFFFAOYSA-N | |
| 3085186 | |
| [H+].[Br-].CC(C)(C)N |
| C4H12BrN | |
| MFCD00674188 | |
| tert-Butylammonium Bromide | |
| hydrogen 2-methylpropan-2-amine bromide |
Specifications
| 60469-70-7 | |
| C4H12BrN | |
| 1 g | |
| CQKAPARXKPTKBK-UHFFFAOYSA-N | |
| hydrogen 2-methylpropan-2-amine bromide | |
| 3085186 | |
| ≥98.0% (N,T) | |
| tert-Butylamine Hydrobromide |
| White-Yellow | |
| MFCD00674188 | |
| tert-Butylammonium Bromide | |
| [H+].[Br-].CC(C)(C)N | |
| 154.05 | |
| 154.05 | |
| Crystalline Powder |
Safety and Handling
TSCA : No