missing translation for 'onlineSavingsMsg'
Learn More

Sigma Aldrich tert-Butylamine

Catalog No. B89205100ML
Encompass
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B89205100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B89205100ML Supplier Sigma Aldrich Supplier No. B89205100ML
Only null left

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

tert-Butylamine

Specifications

CAS 75-64-9
Melting Point -67°C (lit.)
Density 0.696 g/mL (at 25°C (literature))
Boiling Point 46°C (lit.)
Molecular Formula C4H11N
Refractive Index n20/D 1.377 (literature)
Linear Formula (CH3)3 CNH2
Quantity 100 mL
MDL Number MFCD00008050
Synonym TBA; 2-Amino-2-methylpropane
Molecular Weight (g/mol) 73.14
Percent Purity 98%
Show More Show Less