missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sigma Aldrich tert-Butylamine
Description
tert-Butylamine
Specifications
Specifications
| CAS | 75-64-9 |
| Melting Point | -67°C (lit.) |
| Density | 0.696 g/mL (at 25°C (literature)) |
| Boiling Point | 46°C (lit.) |
| Molecular Formula | C4H11N |
| Refractive Index | n20/D 1.377 (literature) |
| Linear Formula | (CH3)3 CNH2 |
| MDL Number | MFCD00008050 |
| Quantity | 1 L |
| Synonym | 2-Amino-2-methylpropane |
| Show More |