missing translation for 'onlineSavingsMsg'
Learn More

tert-Butylacetyl chloride, 98+%

Catalog No. p-7080580
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1016214 25 g
AAA1016222 100 g
AAA1016236 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1016214 Supplier Thermo Scientific Chemicals Supplier No. A1016214
Only null left

CAS: 7065-46-5 | C6H11ClO | 134.603 g/mol

tert-Butylacetyl chloride is used an acyl chloride compound for proteomics research. It is also used as intermediate for important acylation reagents agrochemical and pharmaceutical.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
tert-Butylacetyl chloride is used an acyl chloride compound for proteomics research. It is also used as intermediate for important acylation reagents agrochemical and pharmaceutical.

Solubility
Reacts with water.

Notes
Moisture Sensitive Store away from strong oxidizing agents. keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Chemical Identifiers

CAS 7065-46-5
Molecular Formula C6H11ClO
Molecular Weight (g/mol) 134.603
MDL Number MFCD00000737
InChI Key BUTKIHRNYUEGKB-UHFFFAOYSA-N
Synonym 3,3-dimethylbutyryl chloride, tert-butylacetyl chloride, t-butylacetyl chloride, butanoyl chloride, 3,3-dimethyl, 3,3-dimethyl butyryl chloride, tert-butyl acetyl chloride, neohexanoyl chloride, t-butylacetylchloride, tertbutylacetylchloride, pubchem2093
PubChem CID 81514
IUPAC Name 3,3-dimethylbutanoyl chloride
SMILES CC(C)(C)CC(=O)Cl

Specifications

Density 0.967
Boiling Point 128°C to 130°C
Flash Point 21°C (69°F)
Odor Acrid
Refractive Index 1.423
Quantity 25 g
UN Number UN2924
Beilstein 1740421
Sensitivity Moisture sensitive
Solubility Information Reacts with water.
Formula Weight 134.61
Percent Purity ≥98%
Chemical Name or Material tert-Butylacetyl chloride
Show More Show Less
Hazard Category H225-H314-H335
Hazard Statement GHS H Statement
H225-H314-H318
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, CORROSIVE, N.O.S.
EINECSNumber 230-356-4
TSCA Yes
Recommended Storage Keep cold; Store under Nitrogen

RUO – Research Use Only