missing translation for 'onlineSavingsMsg'
Learn More

tert-Butylacetic Acid 98.0+%, TCI America™

Catalog No. p-7118693
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B082625ML 25 mL
B0826500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B082625ML Supplier TCI America Supplier No. B082625ML

Chemical Identifiers

CAS 1070-83-3
Molecular Formula C6H12O2
Molecular Weight (g/mol) 116.16
MDL Number MFCD00002715
InChI Key MLMQPDHYNJCQAO-UHFFFAOYSA-N
Synonym 3,3-dimethylbutyric acid, tert-butylacetic acid, t-butylacetic acid, butanoic acid, 3,3-dimethyl, 3,3-dimethyl-butanoic acid, unii-q80k0tzq55, butyric acid, 3,3-dimethyl, 3,3-dimethyl-butyric acid, 3,3-dimethyl-n-butyric acid, tert-butyric acid
PubChem CID 14057
ChEBI CHEBI:38647
IUPAC Name 3,3-dimethylbutanoic acid
SMILES CC(C)(C)CC(=O)O

Specifications

Melting Point -11°C
Color Yellow
Boiling Point 190°C
Quantity 25 mL
UN Number 3265
Formula Weight 116.16
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material tert-Butylacetic Acid
EINECSNumber (2)-0608
TSCA Yes