missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl vinyl ether, 98%, stab. with ca 0.1% N,N-diethylaniline

Catalog No. AAH3077006
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3077006 5 g
AAH3077014 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3077006 Supplier Thermo Scientific Chemicals Supplier No. H3077006
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 926-02-3 | C6H12O | 100.16 g/mol

tert-Butyl vinyl ether is used in the synthesis of 3-tert-Butoxycyclobutanone by reacting with ketene using zinc(II) chloride as a catalyst. It acts as an intermediate for the functional vinyl monomer.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
tert-Butyl vinyl ether is used in the synthesis of 3-tert-Butoxycyclobutanone by reacting with ketene using zinc(II) chloride as a catalyst. It acts as an intermediate for the functional vinyl monomer.

Solubility
Miscible with alcohol, carbon disulfide, chloroform and diethyl ether. Slightly miscible with water.

Notes
Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 926-02-3
Molecular Formula C6H12O
Molecular Weight (g/mol) 100.16
MDL Number MFCD00048246
InChI Key PGYJSURPYAAOMM-UHFFFAOYSA-N
Synonym tert-butyl vinyl ether, 2-ethenyloxy-2-methylpropane, propane, 2-ethenyloxy-2-methyl, propane,2-ethenyloxy-2-methyl, t-butyl vinyl ether, tert-butylvinyl ether, vinyltertiary-butylether, tertiarybutyl vinyl ether, vinyl tert.-butyl ether, tertiary butyl vinyl ether
PubChem CID 70220
IUPAC Name 2-ethenoxy-2-methylpropane
SMILES CC(C)(C)OC=C

Specifications

Density 0.762
Boiling Point 75°C to 76°C
Flash Point −17°C (1°F)
Odor Characteristic
Refractive Index 1.399
Linear Formula (CH3)3COCH=CH2
Quantity 5 g
UN Number UN3271
Solubility Information Miscible with alcohol,carbon disulfide,chloroform and diethyl ether. Slightly miscible with water.
Formula Weight 100.16
Percent Purity 98%
Chemical Name or Material tert-Butyl vinyl ether, stabilized with ≈0.1% N,N-diethylaniline
Show More Show Less
Hazard Category H225-H315-H319
Hazard Statement GHS H Statement
H225
Highly flammable liquid and vapor.
Precautionary Statement P210-P233-P240-P241-P242-P243-P264b-P280-P303+P361+P353-P305+P351+P338-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ETHERS, N.O.S.
EINECSNumber 618-860-6
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only