missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl (S)-1,2,3,4-tetrahydroisoquinoline-3-carboxylate hydrochloride, 98%

CAS: 77497-74-6 | C14H19NO2 | 233.311 g/mol

Supplier:  Thermo Scientific Chemicals H6446806

 View more versions of this product

Catalog No. AAH6446806


Only null left
Explore available promotions

Description

Description

It is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as a pharmaceutical intermediate.

Solubility
Soluble in water.

Notes
Store away from heat and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Keep it refrigerated.
Specifications

Specifications

tert-Butyl (S)-1,2,3,4-tetrahydroisoquinoline-3-carboxylate hydrochloride
C14H19NO2
5 g
s-tert-butyl 1,2,3,4-tetrahydroisoquinoline-3-carboxylate, s-1,2,3,4-tetrahydro-3-isoquinolinecarboxylic acid t-butyl ester, tert-butyl 3s-1,2,3,4-tetrahydroisoquinoline-3-carboxylate, tert-butyl s-1,2,3,4-tetrahydro-isoquinoline-3-carboxylate, s-1,2,3,4-tetrahydro-3-isoquinolinecarboxylic acid tert-butyl ester, tic-obut, s-1,2,3,4-tetrahydro-3-isoquinolinecarboxylic ac, tert-butyl s-1,2,3,4-tetrahydroisoquinoline-3-carboxylate, tert.-butyl 3s-1,2,3,4-tetrahydroisoquinoline-3-carboxylate, s-1,2,3,4-tetrahydro-3-isoquinolinecarboxylic acid tert-butyl ester hydrochloride
KRSUDBIZXKOKGA-LBPRGKRZSA-N
tert-butyl (3S)-1,2,3,4-tetrahydroisoquinoline-3-carboxylate
9209323
99%
77497-74-6
MFCD04114841
5744730
Soluble in water.
CC(C)(C)OC(=O)C1CC2=CC=CC=C2CN1
233.311
269.77
Solid

Chemical Identifiers

77497-74-6
233.311
KRSUDBIZXKOKGA-LBPRGKRZSA-N
9209323
CC(C)(C)OC(=O)C1CC2=CC=CC=C2CN1
C14H19NO2
MFCD04114841
s-tert-butyl 1,2,3,4-tetrahydroisoquinoline-3-carboxylate, s-1,2,3,4-tetrahydro-3-isoquinolinecarboxylic acid t-butyl ester, tert-butyl 3s-1,2,3,4-tetrahydroisoquinoline-3-carboxylate, tert-butyl s-1,2,3,4-tetrahydro-isoquinoline-3-carboxylate, s-1,2,3,4-tetrahydro-3-isoquinolinecarboxylic acid tert-butyl ester, tic-obut, s-1,2,3,4-tetrahydro-3-isoquinolinecarboxylic ac, tert-butyl s-1,2,3,4-tetrahydroisoquinoline-3-carboxylate, tert.-butyl 3s-1,2,3,4-tetrahydroisoquinoline-3-carboxylate, s-1,2,3,4-tetrahydro-3-isoquinolinecarboxylic acid tert-butyl ester hydrochloride
tert-butyl (3S)-1,2,3,4-tetrahydroisoquinoline-3-carboxylate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

storageNote1 : 2° to 8°C

Recommended Storage : Keep cold

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only